Masterflex® Adapter Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Supplier: VWR International
MFLX31201-27 MFLX31201-28 MFLX31201-29 MFLX31201-30 MFLX31201-33 MFLX31201-34 MFLX31201-35
MFLX31201-27BG 312.84 CAD
MFLX31201-27 MFLX31201-28 MFLX31201-29 MFLX31201-30 MFLX31201-33 MFLX31201-34 MFLX31201-35
Masterflex® Adapter Fittings, Sanitary Clamp to Barb with TPE Fused Gasket, Avantor®
Fittings and Connectors
Sanitary fitting with built-in gasket makes sanitary installation easier than ever.
  • Sanitary gasket combined with a sanitary fitting adds another layer of safety to installation
  • Prevent contamination with zero user contact with the gasket to ensure sterility
  • Eliminate the risk of misalignment with the limited slip design
Allowing for easy assembly and disassembly of sanitary set-ups, these adapters are suitable for use connecting a Tri-Clamp® fitting to flexible tubing with the hose barb adapter. These sanitary fittings feature a built-in elastomericMore Product Information