Masterflex® L/S® Pump Power Supplies, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
7525-90
MFLX07525-90EA 303.36 CAD
MFLX07525-90
Masterflex® L/S® Pump Power Supplies, Avantor®
Power Supplies
Optimize performance of your Masterflex pump.
  • Suitable for power requirements 90 to 260 VAC
This replacement power supply is suitable for use with the Masterflex C/L Digital pump.