Masterflex® Strainer Gasket for Low-Cost In-Line Strainer Systems, Avantor®
Supplier: SPECIALTY MFG CO
5580710 (11337) 5580300 (10498) 5580480 (11302) 5580080 (10515) 5580730 (11306) 5580380 (10539)
MFLX29595-81EA 11.69 CAD
MFLX29595-81 MFLX29595-83 MFLX29595-85 MFLX29595-87 MFLX29595-89 MFLX29595-91
Masterflex® Strainer Gasket for Low-Cost In-Line Strainer Systems, Avantor®
Filters Strainers
Easily customize strainer system to suit your application.
  • Choose from either EDPM or Viton® gaskets based on chemical compatibility requirements
Build your own economical filtration system to filter out particulates from your fluids. Available in three classes: low-profile, intermediate, and large. Modify or change components of the same class to meet the demands of your application.
Low-Profile Strainers are ideal for small-scale low-volume filtration. Use in research applications, cleaning equipment, and laboratory equipment.
Intermediate StrainersMore Product Information