Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
ACRZB0V19 ACRZS0V12 ACRYS0V05 ACRZB1V05 ACRZB1V19 ACRYB1V19 ACRYB1V26 ACRZS1V05 ACRZS1V12 ACRZS1V19 ACRYS1V12 ACRZB0V05 ACRZB0V12 ACRYB0V19 ACRYB0V26 ACRZS0V05 ACRZS0V19 ACRYS0V12 ACRYS0V26 ACRZB1V12 ACRYB1V05 ACRYB1V12 ACRYS1V05 ACRYS1V19 ACRYS1V24
MFLX68561-44EA 109.02 CAD
MFLX68561-44 MFLX68561-50 MFLX68561-52 MFLX68561-56 MFLX68561-58 MFLX68561-61 MFLX68561-62 MFLX68561-63 MFLX68561-64 MFLX68561-65 MFLX68561-67 MFLX68561-42 MFLX68561-43 MFLX68561-47 MFLX68561-48 MFLX68561-49 MFLX68561-51 MFLX68561-53 MFLX68561-55 MFLX68561-57 MFLX68561-59 MFLX68561-60 MFLX68561-66 MFLX68561-68 MFLX68561-69
Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for helium. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.