Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PMR1-010701 PMR1-010720 PMR1-010722 PMR1-010741 PMR1-010742 PMR1-010745 PMR1-011958 PMR1-010765 P04/1-N034-10ST PMR1-010788 PMR1-010805 PMR1-010808 PMR1-010809 PMR1-010810 PMR1-010574 PMR1-010600 P04/1-044-15ST PMR1-010613 PMR1-010639 PMR1-010640 MFLX32000-28 MFLX32000-38 MFLX32001-28 MFLX32001-32 MFLX32001-36 MFLX32001-38 MFLX32002-34 MFLX32002-36 MFLX32003-30 MFLX32003-32 MFLX32003-34 MFLX32004-34 MFLX32004-36 MFLX32005-30 MFLX32005-32
MFLX32000-34EA 323.9 CAD
MFLX32000-34 MFLX32001-30 MFLX32001-34 MFLX32002-28 MFLX32002-30 MFLX32002-38 MFLX32003-28 MFLX32003-36 MFLX32003-38 MFLX32004-38 MFLX32005-28 MFLX32005-34 MFLX32005-36 MFLX32005-38 MFLX32030-40 MFLX32032-40 MFLX32033-40 MFLX32033-42 MFLX32035-40 MFLX32035-42 MFLX32000-28 MFLX32000-38 MFLX32001-28 MFLX32001-32 MFLX32001-36 MFLX32001-38 MFLX32002-34 MFLX32002-36 MFLX32003-30 MFLX32003-32 MFLX32003-34 MFLX32004-34 MFLX32004-36 MFLX32005-30 MFLX32005-32
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Water, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.