Masterflex® Doppler Ultrasonic Flowmeters, Avantor®
Supplier: BADGER METER INC
DDFXD3-ANNA-NN DT94-020-N000-N DT95-020-N000-N MFLX32615-62 MFLX32615-64
MFLX32615-60EA 4218.6 CAD
MFLX32615-60 MFLX32615-66 MFLX32615-68 MFLX32615-62 MFLX32615-64
Masterflex® Doppler Ultrasonic Flowmeters, Avantor®
Flow Meters
In-field linearization allows adaptation to a wide variety of applications.
  • Accuracy is ±2% full-scale over calibrated span
  • Nonintrusive, clamp-on transducers fit pipes from ¼” and above
  • Clamp-on sensor eliminates the need for in-line flanges, pipe fittings, strainers, or filters
Use these doppler flowmeters measure the flow of liquids in pipes from 1/4” and larger. Each flowmeter requires a transducer (32615-66 or -68) and a monitor (32615-60, -62, or -64).|The transducers send flow data via RG59 coax cable to the monitor, and the monitor’s softwareMore Product Information