Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 65-mm, Avantor®
Supplier: VWR International
MFLX32044-36 MFLX32044-38 MFLX32044-40 MFLX32046-39
MFLX32044-36EA 369.72 CAD
MFLX32044-36 MFLX32044-38 MFLX32044-40 MFLX32046-39
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, High-Flow, 65-mm, Avantor®
Flow Meters
Read more easily with 16% magnification of scale.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).