Masterflex® Direct-Read Variable-Area Flowmeters without Valve, Metric Units, Avantor®
Supplier: KEY INSTRUMENTS, LLC
2510A2A12BNBN-CP 2510A2A13BNBN-CP 2510A2A14BNBN-CP 2510A2A15BNBN-CP 2510A2A17BNBN-CP 2510A2A18BNBN-CP 2520A4L38BNBN-CP 2520A4L56BNBN-CP 2520A4L50BNBN-CP 2520A4L52BNBN-CP 2520A4L53BNBN-CP 2520A4L54BNBN-CP 2530A4L69BNBN-CP 2530A4L71BNBN-CP 2520A4A40BNBN-CP 2520A4A41BNBN-CP 2520A4A42BNBN-CP 2520A4A43BNBN-CP 2520A4A44BNBN-CP 2520A4A47BNBN-CP 2530A4A72BNBN-CP 2530A4A70BNBN-CP 2530A4A73BNBN-CP 2540A5L59PS-CP 2540A4A53PS-CP 2540A4A54PS-CP 2540A5A55PS-CP 2510A2L10BNBN-CP 2510A2L11BNBN-CP 2510A2A16BNBN-CP 2520A4L51BNBN-CP 2540A4L61PS-CP
MFLX32457-40EA 181.7 CAD
MFLX32457-40 MFLX32457-42 MFLX32457-44 MFLX32457-46 MFLX32457-50 MFLX32457-52 MFLX32458-30 MFLX32458-32 MFLX32458-36 MFLX32458-38 MFLX32458-40 MFLX32458-42 MFLX32458-44 MFLX32458-46 MFLX32458-50 MFLX32458-52 MFLX32458-54 MFLX32458-56 MFLX32458-58 MFLX32458-60 MFLX32458-62 MFLX32458-64 MFLX32458-65 MFLX32462-20 MFLX32462-30 MFLX32462-32 MFLX32462-34 MFLX32457-32 MFLX32457-34 MFLX32457-48 MFLX32458-34 MFLX32462-22 MFLX32462-24
Masterflex® Direct-Read Variable-Area Flowmeters without Valve, Metric Units, Avantor®
Flow Meters
Durable single-piece body withstands severe shock and vibration.
  • Ideal for process plant applications on air sampling equipment, gas analyzers and chemical feed systems for water treatment
  • Flexible design allows for panel or bench mounting
  • Variable Area with Direct Metric-Reading Scales
Machined from solid acrylic blocks, these meters have integral metering tubes that provide precise readings even in aggressive plant environments. The meters’ inlet/outlet ports and mounting studs are extended for easy panel installation. An alternate option is a tripod base (sold separately below) which allows for mobility from bench to bench.
Note: There are many additional types of acrylic flowmeters not listed here. Contact our Application Specialists for quotes on acrylic meters with special requirements.