Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, 65-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
P01/6-NO42-07G P01/6-NO42-07ST P01/6-NO32-15ST P01/6-NO22-13G P01/6-NO12-10G P01/6-NO12-10ST P01/6-NO14-96G P01/6-NO14-96ST P01/6-NO54-17ST P11/6-N052-01G P11/6-N0365ST P21/6-N042-07G P21/6-N042-07ST P21/6-N012-10G P21/6-N012-10ST P21/6-N052-01G P21/6-N013-88ST P21/6-N054-17ST MFLX32044-04 MFLX32044-10 MFLX32044-16 MFLX32044-18 MFLX32044-20 MFLX32044-22 MFLX32044-24 MFLX32044-26 MFLX32044-32 MFLX32045-00 MFLX32045-02 MFLX32045-04 MFLX32045-06 MFLX32045-08 MFLX32045-10 MFLX32045-12 MFLX32045-14 MFLX32045-17 MFLX32045-18 MFLX32045-20 MFLX32045-32 MFLX32046-05 MFLX32046-07 MFLX32046-09 MFLX32046-11 MFLX32046-18 MFLX32046-19 MFLX32046-21 MFLX32046-31 MFLX32046-33 MFLX32044-15
MFLX32044-00EA 369.72 CAD
MFLX32044-00 MFLX32044-02 MFLX32044-06 MFLX32044-08 MFLX32044-12 MFLX32044-14 MFLX32044-28 MFLX32044-30 MFLX32044-34 MFLX32045-15 MFLX32045-26 MFLX32046-01 MFLX32046-03 MFLX32046-13 MFLX32046-15 MFLX32046-16 MFLX32046-23 MFLX32046-35 MFLX32044-04 MFLX32044-10 MFLX32044-16 MFLX32044-18 MFLX32044-20 MFLX32044-22 MFLX32044-24 MFLX32044-26 MFLX32044-32 MFLX32045-00 MFLX32045-02 MFLX32045-04 MFLX32045-06 MFLX32045-08 MFLX32045-10 MFLX32045-12 MFLX32045-14 MFLX32045-17 MFLX32045-18 MFLX32045-20 MFLX32045-32 MFLX32046-05 MFLX32046-07 MFLX32046-09 MFLX32046-11 MFLX32046-18 MFLX32046-19 MFLX32046-21 MFLX32046-31 MFLX32046-33 MFLX32044-15
Masterflex® Correlated Variable-Area Flowmeters, High-Res Valve, 65-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).