Masterflex® Ultra-High Pressure Dual Piston Pumps, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CP024SCL
MFLX74931-40EA 18122.6 CAD
MFLX74931-40
Masterflex® Ultra-High Pressure Dual Piston Pumps, Avantor®
Pumps Piston Pumps
No more sloppy chromatograms.
  • Flow rates from 5 to 12 ml/min
  • Operate pump via keypad, PC, or use voltage flow and digital run/stop controls
  • Automatic shutoff when pressure exceeds upper or lower limits prevents damage from malfunctions
  • Digital stepper motor ensures accuracy at low flow
Constant pressure model (74931-40) is perfect for column packing. Advanced pressure control algorithms ensure consistent bed density. Selectable process parameters include pressure set point, ramp rate, and upper flow rate limit.
Delivery information: Includes removable drip tray with leak sensor and detachable power cord.