Masterflex® Pressure Booster Pump Systems, Avantor®
Supplier: VWR International
MFLX70794-04 MFLX70794-22 MFLX70794-24 MFLX70794-44 MFLX70794-64
MFLX70794-04EA 6383.2 CAD
MFLX70794-04 MFLX70794-22 MFLX70794-24 MFLX70794-44 MFLX70794-64
Masterflex® Pressure Booster Pump Systems, Avantor®
Pumps Piston Pumps
Pump system automatically adjusts speed to maintain set pressure.
  • Maximum flow range of 25 to 100 GPM│NEMA 1 or NEMA 4 variable frequency drive controller with corresponding main disconnect
  • Three-phase input line voltage (single-phase systems are available)
Pump supply booster systems feature a stainless steel centrifugal pump. System is controlled by the variable frequency drive (VFD) which features an LCD and adjustable pressure range; choose the VFD with either a NEMA 1 or NEMA 4 rated enclosure and main disconnect.
System has a small footprint of approximately 30" x 30" (76.2 x 76.2 cm), and has a 1.5" NPT(F) suction and discharge connection. Piping, valve, and drain bodies are made from brass to meet most needs.
Additional options are available: stainless steel piping upgrade, suction check valve (bronze), unions (bronze), low suction shutoff, and low suction shutoff with alarm. If your facility has only single phase voltage, a custom unit can be made to match. Please contact our Application Specialists for details.
Please contact your local Avantor office to customize a system suited to your needs.
Delivery information: Includes pump, VFD controller, bladder tank, suction and discharge gauges, discharge check valve, backup relief valve, and necessary piping and drain.