Masterflex® Submersible Dewatering Centrifugal Pumps, Avantor®
Supplier: VWR International
EPD-3AS1 EPD-3MS1 - 7550-74 EPD-7MT4 OPTIMA-3AS1-SLIMFLT
MFLX75504-50EA 875.32 CAD
MFLX75504-50 MFLX75504-60 MFLX75507-60 MFLX75507-72 MFLX75507-50 MFLX75507-52 MFLX75507-70 MFLX75507-74 MFLX75507-76 MFLX75504-55
Masterflex® Submersible Dewatering Centrifugal Pumps, Avantor®
Pumps Centrifugal Pumps
Semi-open impeller design pumps solids up to 3/8" in slurries and up to 81 GPM.
  • Stainless steel construction resists corrosion and rust
  • Pumps with automatic operation have float switch; manual operation models activate with a switch
  • Air-filled motors have permanent split capacitor design with built-in thermal protection and automatic reset
Operate pumps either fully or partially submerged. Pumps are ideal for slurries and dirty or semi-dirty waters.
Delivery information: Includes 20 ft (6.1 m) water-resistant #16 AWG cord.