Masterflex® Ismatec® Reglo Digital Gear Pump Drives with MasterflexLive®, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
78018-50 78018-50 78018-50 78018-50 78018-50 78018-50 78018-50 78018-50 78018-50
MFLX78018-50-EUCS 0 CAD
MFLX78018-50-EU MFLX78018-50-IT MFLX78018-50-UK MFLX78018-50-DK MFLX78018-50 MFLX78018-50-IL MFLX78018-50-AU MFLX78018-50-CH MFLX78018-50-IN
Masterflex® Ismatec® Reglo Digital Gear Pump Drives with MasterflexLive®, Avantor®
Pumps Pump Drives Gear Pump Drives
Achieve high pressure and accurate, pulseless flow -- with an intuitive touch-screen interface.
  • Compact, benchtop pumps optimize ease of use, accuracy, and reliability
  • Compatible with Micropump A-mount and B-mount style gear pump heads for flow ranges from 1.7 to 4680 ml/min
  • 5” (12.7 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
  • Flexible operational control via screen or analog remote control
  • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
  • Pump lets you save and retrieve commonly used dispense programs for rapid set up
  • Speed ramping available in Volume dispense mode – ensure accuracy and minimize sample loss
  • Three-level user access – gives you the option of controlling who can operate the pump and at what level
MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters – including speed, flow rate, dispense volume, and more. You can monitor critical processes running 24/7 without the need for onsite personnel while push notifications alert you to operating conditions and error messages.
Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive; USB connection allows for downloadable firmware updates to be loaded into the pump.
Ordering information: Each model includes a country specific plug/cord set; choose the catalog number with desired country plug type. All units include an IEC 320 socket with a detachable standard US three prong power cable. See country plug description https://us.vwr.com/cms/tech-article-ac-power-plugs-sockets-universal-pattern-guide.
Delivery information: Includes pump drive, B-mount style pump head adapter, and power adapter with 1.8 m (6 ft.) line cord. These models are shipped with country specific plug/cord set; please order the specific catalog number with the desired plug type.