Masterflex® L/S® Precision Process Drives, Avantor®
Supplier: Avantor Fluid Handling
7528-40 7528-50 7528-40 MFLX07528-45-AU 7528-50 7528-40 MFLX07528-45-DK 7528-50 7528-40 MFLX07528-45-IN 7528-50 7528-40 MFLX07528-45-IL 7528-50 7528-40 MFLX07528-45-EU 7528-50 7528-40 MFLX07528-45-UK 7528-50 7528-40 MFLX07528-45-CH 7528-50 7528-40 MFLX07528-45-IT 7528-50 MFLX07528-55-AU MFLX07528-55-DK MFLX07528-55-IN MFLX07528-55-IL MFLX07528-55-EU MFLX07528-55-UK MFLX07528-55-CH MFLX07528-55-IT
MFLX07528-40EA 7078.4 CAD
MFLX07528-40 MFLX07528-45 MFLX07528-50 MFLX07528-40-AU MFLX07528-45-AU MFLX07528-50-AU MFLX07528-40-DK MFLX07528-45-DK MFLX07528-50-DK MFLX07528-40-IN MFLX07528-45-IN MFLX07528-50-IN MFLX07528-40-IL MFLX07528-45-IL MFLX07528-50-IL MFLX07528-40-EU MFLX07528-45-EU MFLX07528-50-EU MFLX07528-40-UK MFLX07528-45-UK MFLX07528-50-UK MFLX07528-40-CH MFLX07528-45-CH MFLX07528-50-CH MFLX07528-40-IT MFLX07528-45-IT MFLX07528-50-IT MFLX07528-55 MFLX07528-55-AU MFLX07528-55-DK MFLX07528-55-IN MFLX07528-55-IL MFLX07528-55-EU MFLX07528-55-UK MFLX07528-55-CH MFLX07528-55-IT
Masterflex® L/S® Precision Process Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Flow rate 0.06 to 3400 ml/min (depends on drive rpm and tubing size)
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for multichannel applications
  • Separate speed and power (on/off) control—maintain speed setting while turning pump on/off
  • NEMA 4X (IP66) rated housing
  • Remote control capabilities via a watertight connector on back of drive
These accurate console drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications. Remote speed control requires cable assembly 77300-32. Operation of the integrated open-head sensor requires use of sensor-equipped L/S Easy-Load II pump heads 77200-30 or 77200-32.
Delivery information: Includes 6-ft (1.8-m) line cord with IEC 320/CEE22 socket. These models are shipped with country-specific plug/cord set; please specify ultimate destination when ordering.