Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
Supplier: Avantor Fluid Handling
7559-00 7559-05 7559-10 7559-15
MFLX07559-00EA 2496.4 CAD
MFLX07559-00 MFLX07559-05 MFLX07559-10 MFLX07559-15
Masterflex® L/S® Variable-Speed Economy Modular Drives, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Versatile variable-speed drive with a wide flow range and simple controls.
  • Flow rate 0.06 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Simple operation with single-turn speed control
  • Separate motor and controller are connected via a 6-ft (1.9 m) cable — place components where convenient or where space allows
  • Single-turn speed control and on/off switch with green power indicator — maintain speed setting while turning pump on/off
  • The 1/10-hp reversible motor has soft start and back EMF for ±2% speed control
More Product Information