Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Supplier: Avantor Fluid Handling
7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10 7594-00 7594-10
MFLX07594-00EA 9464.2 CAD
MFLX07594-00 MFLX07594-10 MFLX07594-00-AU MFLX07594-10-AU MFLX07594-00-DK MFLX07594-10-DK MFLX07594-00-IN MFLX07594-10-IN MFLX07594-00-IL MFLX07594-10-IL MFLX07594-00-EU MFLX07594-10-EU MFLX07594-00-UK MFLX07594-10-UK MFLX07594-00-CH MFLX07594-10-CH MFLX07594-00-IT MFLX07594-10-IT
Masterflex® I/P® Digital Modular Drives with Remote Input/Output, Avantor®
Pumps Pump Drives Peristaltic Pump Drives
Place motor and controller where convenient with modular design.
  • Flow rate 0.0006 to 26 LPM with I/P tubing—flow rate depends on drive rpm, pump head, number of pump heads, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Select your preferred language from the user-friendly menu for programming and operation
  • Dispense by Volume in ml (0.001 to 9,999 mL), L,
More Product Information