Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Supplier: Avantor Fluid Handling
77220-20 77220-30
MFLX77220-20EA 462.94 CAD
MFLX77220-20 MFLX77220-30
Masterflex® L/S® MiniFlex® Pump Heads, Avantor®
Pumps Pump Heads Peristaltic Pump Heads
Pump head accepts continuous lengths of tubing for clean flow paths.
  • Flow range of 1.8 to 220 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Dual-channel model delivers synchronous flow from two separate channels; single-channel model delivers highest flow rate
  • Automatic tubing retention makes tube loading quick and easy
  • Suitable for use with Masterflex® L/S® Digital Miniflex® Pump Systems
The Masterflex® Miniflex® pump heads feature a wide occlusion angle that prevents fluid backflow up to the rated pressure of the tubing. Self-lubricating thermoplastic polyester rollers are low friction and long wearing for smooth operation and long life. A compact overall size envelope makes these heads ideal for OEM applications and integration into fluid systems and machinery.