Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pumps, Avantor®
Supplier: VWR International
MFLX74905-19 MFLX74905-39
MFLX74905-19EA 9432.6 CAD
MFLX74905-19 MFLX74905-39
Masterflex® Touch-Screen, Continuous-Cycle, Four-Syringe Pumps, Avantor®
Pumps Syringe Pumps
Continuously cycle up to four syringes in a push-pull action.
  • Flow range from 5 pL/min to 215.8 ml/min
  • Intuitive touch screen allows you to quickly create and recall configurations
  • You can daisy chain up to 99 pumps together using the built in RS-485
  • Accuracy of ±0.35% and reproducibility of ±0.05%
Hold up to four syringes to cycle continuously back and forth in a push-pull action. As two syringes are infusing, two syringes are simultaneously withdrawing at the same rate. At the end of the set volume the direction is automatically reversed and the n
Delivery information: Includes 6 ft (1.8 m) U.S. power cord.