Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40 7525-20 7525-40
MFLX07525-20CS 0 CAD
MFLX07525-20 MFLX07525-40 MFLX07525-20-AU MFLX07525-40-AU MFLX07525-20-DK MFLX07525-40-DK MFLX07525-20-IN MFLX07525-40-IN MFLX07525-20-IL MFLX07525-40-IL MFLX07525-20-EU MFLX07525-40-EU MFLX07525-20-UK MFLX07525-40-UK MFLX07525-20-CH MFLX07525-40-CH MFLX07525-20-IT MFLX07525-40-IT
Masterflex® L/S® Digital Miniflex® Pump Systems, Avantor®
Pumps Peristaltic Pumps
Make low-flow pumping easy!
  • Flow range 0.004 to 315 ml/min—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode
  • Remote control available using analog communication—voltage, current and contact closure
  • Integrated “open-head” sensor halts operation when pump head is opened
  • Pump head accepts continuous lengths of tubing for clean flow paths
  • Automatic tubing retention makes tube loading quick and easy
  • More than 20% smaller than competitive units—compact size and light weight
More Product Information