Masterflex® I/P® Digital Modular Drive Systems, Avantor®
Supplier: Avantor Fluid Handling
77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30 77962-20 77962-30
MFLX77962-20EA 8769 CAD
MFLX77962-20 MFLX77962-30 MFLX77962-20-AU MFLX77962-30-AU MFLX77962-20-DK MFLX77962-30-DK MFLX77962-20-IN MFLX77962-30-IN MFLX77962-20-IL MFLX77962-30-IL MFLX77962-20-EU MFLX77962-30-EU MFLX77962-20-UK MFLX77962-30-UK MFLX77962-20-CH MFLX77962-30-CH MFLX77962-20-IT MFLX77962-30-IT
Masterflex® I/P® Digital Modular Drive Systems, Avantor®
Pumps Peristaltic Pumps
Place motor and controller where convenient with modular design.
  • Flow rate 0.06 to 8 LPM with I/P tubing—flow rate depends on drive rpm, pump head, and tubing size
  • Digital interface with LED display shows pump rpm— confirm speed setting at a glance
  • Remote control available using analog communication—voltage, current and contact closure
  • Powerful motor lets you stack two pump heads for increased flow rate while offering ±0.25% speed control accuracy
  • Membrane keypad allows for easy up-front control of speed, direction, and start/stop
I/P® modular drivesMore Product Information