Masterflex® L/S® Variable Speed Analog Console Pump Systems, Avantor®
Supplier: Avantor Fluid Handling
77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77916-10 77916-20 77925-10 77926-10 77927-10
MFLX77916-10EA 4424 CAD
MFLX77916-10 MFLX77916-20 MFLX77925-10 MFLX77926-10 MFLX77916-10-AU MFLX77916-20-AU MFLX77925-10-AU MFLX77926-10-AU MFLX77916-10-DK MFLX77916-20-DK MFLX77925-10-DK MFLX77926-10-DK MFLX77916-10-IN MFLX77916-20-IN MFLX77925-10-IN MFLX77926-10-IN MFLX77916-10-IL MFLX77916-20-IL MFLX77925-10-IL MFLX77926-10-IL MFLX77916-10-EU MFLX77916-20-EU MFLX77925-10-EU MFLX77926-10-EU MFLX77916-10-UK MFLX77916-20-UK MFLX77925-10-UK MFLX77926-10-UK MFLX77916-10-CH MFLX77916-20-CH MFLX77925-10-CH MFLX77926-10-CH MFLX77916-10-IT MFLX77916-20-IT MFLX77925-10-IT MFLX77926-10-IT MFLX77927-10
Masterflex® L/S® Variable Speed Analog Console Pump Systems, Avantor®
Pumps Peristaltic Pumps
Don’t lose control of your metering applications—digital display gives you precise control.
  • Accurate and repeatable settings with 3-digit LED display of rpm
  • Pump fluid in either direction with reversible motor—perfect for purging tubing
  • Choose multichannel systems for synchronous flow from all channels
  • Maximize processes in a minimal footprint by stacking splash-resistant, IP33-rated drives
  • Two ways to control speed, direction, and start/stop—use either the membrane keypad or remote-control capabilities
These L/S® precision variable-speed console drives offer precise ±0.25% speed control for accurate flow regulation. The large LED display and LED indicators confirm motor rpm and motor direction at a glance. The complete pump systems include all you need to start pumping.