Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Supplier: Avantor Fluid Handling
7528-82 77097-20 MFLX77097-02
MFLX07528-82EA 66.52 CAD
MFLX07528-82 MFLX77097-20 MFLX77097-02
Masterflex® Open-Head Sensor Interlock Retrofit System Accessories, Avantor®
Pumps Peristaltic Pumps
Make the most of your Masterflex® Open-Head Sensor with these accessories and replacement parts.
  • Use in conjunction with Masterflex® L/S® and I/P® open-head sensor equipped pump heads
  • Compatible with Masterflex® L/S® and I/P® pumps without integrated open-head sensor
  • Controller accepts input signals from up to two pump heads
  • Protect technicians and operators tasked with tubing changes and routine maintenance in busy production environments
  • Controller is NEMA 4X / IP66 rated for washdown
The retrofit system is compatible with all I/P® pumps and with L/More Product Information