Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Supplier: VWR International
MFLX75220-00 MFLX75220-20 MFLX75220-30
MFLX75220-00EA 13840.8 CAD
MFLX75220-00 MFLX75220-20 MFLX75220-30
Masterflex® High-Viscosity Sanitary Drum Pumps, Avantor®
Pumps Drum/Transfer Pumps
Capable of pumping fluids with viscosities up to 10,000 cps.
  • Progressive cavity design allows for low-shear properties
  • Flow rates up to 12 GPM (45 LPM)
  • 316 stainless steel construction with PTFE elastomers
These drum pumps are engineered specifically for transferring food products, pharmaceuticals, and cosmetic products from barrels, kettles, and tanks. They offer a progressive cavity, positive-displacement delivery system resulting in a continuous flow of material with little product degradation. This unique design allows you to quickly dismantle, clean, and re-assemble the pump.
Delivery information: Includes drive and pump tube.