Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Supplier: VWR International
MFLX32772-00 MFLX32772-02 MFLX32772-08 MFLX32772-04 MFLX32772-10 MFLX32772-12 MFLX32772-20 MFLX32772-22 MFLX32772-30 MFLX32772-32 MFLX68964-02
MFLX32772-00EA 455.04 CAD
MFLX32772-00 MFLX32772-02 MFLX32772-08 MFLX32772-04 MFLX32772-10 MFLX32772-12 MFLX32772-20 MFLX32772-22 MFLX32772-30 MFLX32772-32 MFLX68964-02
Masterflex® Electro-Optic Point-Level Sensors, Avantor®
Level Sensors
No clog or wear—contain no moving parts.
  • Detect liquids regardless of conductivity, color, or specific gravity
  • Economical and compact — built-in solid-state switching electronics
  • One-unit installation — simply screws into the top or side of your tank
These dependable, electro-optic level sensors contain no moving parts to clog or wear. Sensors offer ±1 mm repeatability and are available in a wide variety of materials to meet even the most demanding applications.
Installation is simple and quick through the tank top, bottom, or side. The switches can be used for low-, intermediate-, or high-liquid level detection. Solid state switching ensures dependability over a long service life. Order controller separately.