Masterflex® Transfer Tubing, PVC, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
CG-375X625-50 CG-625X875-50 4140
MFLX96605-10EA 178.54 CAD
MFLX96605-10 MFLX96605-25 MFLX96605-15
Masterflex® Transfer Tubing, PVC, Avantor®
Tubing Transfer Tubing
Stop oxidation and bacteria problems.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
This tubing is extremely flexible and transparent and is resistant to oxidation, bacteria and a broad range of chemicals. PVC tubing meets FDA CFR Title 21 177.101.