Masterflex® Ismatec® Pump Tubing, Versilon™ 2001, Avantor®
Supplier: VWR International
MFLX96467-14 MFLX96467-28 MFLX96467-36 MFLX96467-42 MFLX96467-48 MFLX96467-22
MFLX06518-04EA 153.1 CAD
MFLX06518-04 MFLX06518-08 MFLX06518-14 MFLX06518-16 MFLX06518-18 MFLX06518-22 MFLX06518-64 MFLX06518-66 MFLX96467-14 MFLX96467-28 MFLX96467-36 MFLX96467-42 MFLX96467-48 MFLX96467-22
Masterflex® Ismatec® Pump Tubing, Versilon™ 2001, Avantor®
Tubing Peristaltic Pump Tubing
Compatible with aggressive chemicals – even MEK!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Outstanding chemical resistance — withstands acetone, DMSO, ketones, and more
  • Cost-effective alternative to fluoropolymers or fluoroelastomers
The Tygon Chemical 2001 translucent tubing hasMore Product Information