Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Versilon™ 2001, Avantor®
Supplier: VWR International
MFLX96466-14 MFLX96466-22 MFLX96466-28 MFLX96466-36 MFLX96466-42 MFLX96466-48
MFLX96466-14EA 202.24 CAD
MFLX96466-14 MFLX96466-22 MFLX96466-28 MFLX96466-36 MFLX96466-42 MFLX96466-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, Versilon™ 2001, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The Tygon® Chemical 2001 tubing is clear in appearance is temperature resistant from -108 °F to 135 °F, and features excellent chemical compatibility to weak and medium acids and bases, salts, and alcohol.
The overall length of Tygon Chemical® 96446-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.