Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Supplier: VWR International
MFLX95663-09 MFLX95663-10 MFLX95663-12 MFLX95663-14 MFLX95663-18 MFLX95663-22 MFLX95663-24 MFLX95663-26 MFLX95663-28 MFLX95663-30 MFLX95663-32 MFLX95663-34 MFLX95663-36 MFLX95663-38 MFLX95663-40 MFLX95663-42 MFLX95663-44 MFLX95663-48 MFLX95663-49 MFLX95663-13 MFLX95663-46
MFLX95663-09EA 70.94 CAD
MFLX95663-09 MFLX95663-10 MFLX95663-12 MFLX95663-14 MFLX95663-18 MFLX95663-22 MFLX95663-24 MFLX95663-26 MFLX95663-28 MFLX95663-30 MFLX95663-32 MFLX95663-34 MFLX95663-36 MFLX95663-38 MFLX95663-40 MFLX95663-42 MFLX95663-44 MFLX95663-48 MFLX95663-49 MFLX95663-13 MFLX95663-46
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Puri-Clear™ LL, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Supple and strong tubing with high stability pumping life
  • Pthalate-free, USP Class VI PVC
Two-Stop PVC Color-coded Autoanalysis Tubing for Ismatec® Pumps is USP Class VI tubing that isMore Product Information