Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Food, Avantor®
Supplier: VWR International
MFLX96457-12 MFLX96457-18 MFLX96457-26 MFLX96457-30 MFLX96457-34 MFLX96457-42 MFLX96457-48
MFLX96457-12EA 167.48 CAD
MFLX96457-12 MFLX96457-18 MFLX96457-26 MFLX96457-30 MFLX96457-34 MFLX96457-42 MFLX96457-48
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Tygon® S3™ E-Food, Avantor®
Tubing Peristaltic Pump Tubing
Long service life - Outstanding fatigue resistance due to flexing!
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Specially designed for use with food applications
  • Extremely smooth bore
Appearance: Opaque
Temperature: -70 to 52 °C
Life at 100 rpm: 1000 hrs
The first of a series of two-stop extension tubing options is below under Accessories. See all of our two-stop extension tubing formulations or contact our Application Specialists for personal assistance.