Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Versilon™ 2001, Avantor®
Supplier: VWR International
07MHSL-04I 07MHSL-06I 07MHSL-09I 07MHSL-19I 07MHSL-15I 07MHSL-18I
MFLX96465-14EA 191.18 CAD
MFLX96465-14 MFLX96465-22 MFLX96465-28 MFLX96465-36 MFLX96465-42 MFLX96465-48
Masterflex® Ismatec® Pump Tubing, 2-Stop Microbore, Versilon™ 2001, Avantor®
Tubing Peristaltic Pump Tubing
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
The overall length of Tygon Chemical 96465-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 381 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.