Masterflex® B/T® PerfectPosition® Pump Tubing, C-Flex®, Avantor®
Supplier: VWR International
MFLX06424-91 MFLX06424-87 MFLX06424-90
MFLX06424-91EA 390.26 CAD
MFLX06424-91 MFLX06424-87 MFLX06424-90
Masterflex® B/T® PerfectPosition® Pump Tubing, C-Flex®, Avantor®
Tubing Peristaltic Pump Tubing
Ensure optimum performance from your Masterflex® pump.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
  • Heat sealable and weldable
  • Inexpensive
  • Biocompatible
  • Physical properties similar to silicone with chemical compatibility of Tygon®
This tubing Complies complies to USP Class VI, EP, REACH, RoHS, and ADCF requirements. It is also heat sealable, weldable, and economical for a wide range of pumping applications.