Masterflex® I/P® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Supplier: VWR International
MFLX06434-26 MFLX06434-73 MFLX06434-82 MFLX06437-73
MFLX06434-26EA 368.14 CAD
MFLX06434-26 MFLX06434-73 MFLX06434-82 MFLX06437-73
Masterflex® I/P® Precision Pump Tubing, C-Flex® ULTRA, Avantor®
Tubing Peristaltic Pump Tubing
Ultra performance for ultrapure applications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Longest pumping life of any C-Flex® formulation
Masterflex® C-Flex® ULTRA Precision pump tubing features extremely low spallation with a smooth inner surface for reduces residue and maintain fluid purity. This tubing will maintain flexibility over a wide temperature range. Heat sealable and weldable, C-Flex ULTRA tubing is ideal single-use assemblies, pharmaceutical and high purity applications.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.