Masterflex® L/S® Bulk-Packed Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96404-13 MFLX96404-14 MFLX96404-15 MFLX96404-16 MFLX96404-17 MFLX96404-18 MFLX96404-24 MFLX96404-25 MFLX96404-35 MFLX96404-36
MFLX96404-13EA 2528 CAD
MFLX96404-13 MFLX96404-14 MFLX96404-15 MFLX96404-16 MFLX96404-17 MFLX96404-18 MFLX96404-24 MFLX96404-25 MFLX96404-35 MFLX96404-36
Masterflex® L/S® Bulk-Packed Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
Our bulk tubing comes in convenient cases containing up to 20 sealed bags of tubing (depending on tubing size). Bulk tubing eliminates individual cardboard boxes, reduces unpacking time, and saves you money on shipping. Tubing comes in 25-ft bags.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.