Masterflex® L/S® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Supplier: VWR International
MFLX96403-13 MFLX96403-14 MFLX96403-15 MFLX96403-16 MFLX96403-17 MFLX96403-18 MFLX96403-24 MFLX96403-25 MFLX96403-35 MFLX96403-36
MFLX96403-13CS 2022 CAD
MFLX96403-13 MFLX96403-14 MFLX96403-15 MFLX96403-16 MFLX96403-17 MFLX96403-18 MFLX96403-24 MFLX96403-25 MFLX96403-35 MFLX96403-36
Masterflex® L/S® Spooled Pump Tubing, Platinum-Cured Silicone, Avantor®
Tubing Peristaltic Pump Tubing
Excellent biocompatibility that meets multiple classifications.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases and spools for customer-friendly ordering and delivery
  • Slightly greater clarity than peroxide-cured tubing
  • Smooth surfaces; lower protein binding levels
  • Fewer potential leachables
Applications: Pharmaceutical, biotech, peristaltic pumping, and food applications
Characteristics: Excellent heat resistance, low compression set, odorless, nontoxic, translucent
Spooled pump tubing provides continuous lengths of pump tubing on convenient, manageable spools that are easy to transport and require less storage space than the equivalent amount of individual tubing coils.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.