Masterflex® Flanged Sanitary Gaskets, Tri-Clamp®, Avantor®
Supplier: VWR International
MFLX30564-28 MFLX30564-44 MFLX30564-46
MFLX30564-28EA 127.82 CAD
MFLX30564-28 MFLX30564-44 MFLX30564-46
Masterflex® Flanged Sanitary Gaskets, Tri-Clamp®, Avantor®
Seals Gaskets
Hands-free installation for difficult-to-reach locations!
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Single-use custom configuration solutions to minimize downtime
With the same benefits as standard sanitary gaskets, these gaskets offer an outer lip to grip the sanitary coupling port—once in place, no need to worry about maintaining the gasket's placement! An especially useful feature for hard-to-reach or vertical clamp joints, using the outer lip speeds assembly and reduces your line's down time.
Available in platinum-cured silicone, EPDM, Buna N and FKM (see material descriptions under standard sanitary fittings for more information).