Masterflex® Single-Use Tubing Pinch Clamp, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
19999-99
MFLX19999-99EA 71.1 CAD
MFLX19999-99
Masterflex® Single-Use Tubing Pinch Clamp, Avantor®
Clamps Tubing and Sanitary Clamps
Swing-away lockdown nut provides ease of use.
  • Higher tensile strength than polysulfone—less likely to break
  • Offers quick release from locked position
  • Ideal for biopharma applications that require gamma irradiation or autoclave cycles
This clamp accepts tubing from 0.187 to 0.500" inner diameters. The recommended wall thickness is 0.085 to 0.200". Tubing materials that can be used include silicone, TPE TPV, PVC, and other rubbers. The maximum hardness for correct functioning of the clamp is 70 Shore A. May be installed on existing tube assemblies without disrupting the process fluid path.