Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PFA, Avantor®
Supplier: ANDON SPECIALTIES INC.
BC6-2 BC8-4 MFLX06383-94 MFLX06383-90
MFLX06383-91EA 77.42 CAD
MFLX06383-91 MFLX06383-92 MFLX06383-93 MFLX06383-94 MFLX06383-90
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PFA, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.