Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, Nylon, Avantor®
Supplier: VWR International
MFLX30401-05 MFLX30401-07 MFLX30401-21 MFLX30622-85 MFLX30622-87 MFLX30622-90 MFLX30622-94 MFLX30622-97 MFLX30622-99 MFLX30623-01 MFLX30623-03 MFLX30623-07 MFLX30623-09 MFLX30623-11 MFLX30623-13 MFLX30623-15 MFLX30626-87 MFLX31702-28 MFLX30401-14 MFLX30401-17 MFLX30401-19 MFLX30401-24 MFLX30623-05 MFLX30626-97 MFLX30627-15 MFLX30401-00 MFLX30626-94
MFLX30401-05EA 38.55 CAD
MFLX30401-05 MFLX30401-07 MFLX30401-21 MFLX30622-85 MFLX30622-87 MFLX30622-90 MFLX30622-94 MFLX30622-97 MFLX30622-99 MFLX30623-01 MFLX30623-03 MFLX30623-07 MFLX30623-09 MFLX30623-11 MFLX30623-13 MFLX30623-15 MFLX30626-87 MFLX31702-28 MFLX30401-14 MFLX30401-17 MFLX30401-19 MFLX30401-24 MFLX30623-05 MFLX30626-97 MFLX30627-15 MFLX30401-00 MFLX30626-94
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Elbow, Nylon, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed elbowMore Product Information