Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Supplier: VWR International
MFLX30800-16 MFLX30800-18 MFLX30800-20 MFLX30800-22 MFLX30800-24 MFLX30800-26 MFLX30800-28 MFLX45518-00 MFLX45518-02 MFLX45518-06 MFLX45518-08 MFLX45518-11 MFLX45518-22 MFLX45518-24 MFLX45518-26 MFLX45518-44
MFLX30800-16CS 0 CAD
MFLX30800-16 MFLX30800-18 MFLX30800-20 MFLX30800-22 MFLX30800-24 MFLX30800-26 MFLX30800-28 MFLX45518-00 MFLX45518-02 MFLX45518-06 MFLX45518-08 MFLX45518-11 MFLX45518-22 MFLX45518-24 MFLX45518-26 MFLX45518-44
Masterflex® Adapter Fittings, Male Luer to Hose Barb, Straight, Polypropylene, Avantor®
Fittings and Connectors
Easily transition to a luer fitting.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.