Masterflex® One-Way Luer Check Valve Fittings, Luer, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
SCV23050 (10PK) SCV08053
MFLX30505-91CS 0 CAD
MFLX30505-91 MFLX30505-92
Masterflex® One-Way Luer Check Valve Fittings, Luer, Straight, Avantor®
Fittings and Connectors
Get a seamless transition from source to system.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Valves feature low priming volume, ease of removing air bubbles during priming and operate in any spatial orientation. Valves are manufactured DEHP-and latex-free silicone diaphragms. Materials meet USP Class VI and ISO 10993 specifications.