Masterflex® Plug Fittings, Male NPT Threaded, Straight, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
P8NN 10PK P6HDPE 100PK P4HDPE 100PK P12HDPE 10PK P8HDPE 10PK P8HDPE 100PK P4NN 100PK P12NN 100PK P12NN 10PK P4NN 10PK SS5406-2P EA P6NN 10PK P8NN 100PK P4-PP HPM-1800-10 P4HDPE 10PK HPM-1800B-100 P6HDPE 10PK SS5406-4P EA SS5406-8P EA P6NN 100PK P16NN 10PK HPM-1800B-10
MFLX41519-49EA 33.5 CAD
MFLX41519-49 MFLX31701-90 MFLX30401-39 MFLX30401-43 MFLX30401-53 MFLX30401-42 MFLX30401-45 MFLX30401-44 MFLX30401-47 MFLX30401-36 MFLX30401-46 MFLX30401-38 MFLX30401-48 MFLX31411-08 MFLX30401-50 MFLX31411-06 MFLX30401-41 MFLX31701-91 MFLX30401-52 MFLX31411-05 MFLX30401-40 MFLX31701-92 MFLX30401-51
Masterflex® Plug Fittings, Male NPT Threaded, Straight, Avantor®
Fittings and Connectors
Deigned for permanent connection.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Very good chemical resistance. Rated to 200 psi at 77 °F ( 25 °C). Usable from -65 to 190 °F (-53 to 87 °C).