Masterflex® Eccentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
Supplier: VWR International
MFLX31805-01 MFLX40619-93
MFLX31805-01EA 333.38 CAD
MFLX31805-01 MFLX40619-93
Masterflex® Eccentric Reducer Fittings, Sanitary Clamp, Straight, Avantor®
Fittings and Connectors
Maintain sanitary conditions in your processes while decreasing diameter.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
  • All fittings are compatible with Tri-Clamp® fittings
  • Excellent chemical resistance; low leachable characteristics
Designed with polypropylene the fitting performs more consistently after repeated autoclaving. Clamps and gaskets required for a complete system are sold separately.