Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
RT4-2-200NK7-MF 10PK RT12-8HDPE 100PK RT2-4-200NK7-MF 10PK RT10-6NK7-MF 10PK RT12-8HDPE 10PK THB-1412-10 RT8-3-8 EA RT8-6NN 10PK RT1-3-1 EA THB-1438-10 RT8-6HDPE 10PK RT3-1-3 EA THB-3638B-10 RT5-3-5 EA RT6-3-6 EA RT12-6NK7-MF 10PK THB-3638-10 RT10-6HDPE 10PK RT8-6HDPE 100PK THB-1412-100 RT8-6-8 EA RT4-200NK7-MF 10PK RT12-8NN 10PK RT6-8-6 EA
MFLX40615-50EA 115.02 CAD
MFLX40615-50 MFLX31208-47 MFLX30401-54 MFLX31208-46 MFLX30401-56 MFLX30401-55 MFLX31208-48 MFLX30401-58 MFLX40615-51 MFLX31702-47 MFLX31702-57 MFLX30401-59 MFLX31702-45 MFLX31208-41 MFLX40614-36 MFLX40615-48 MFLX31702-62 MFLX31702-61 MFLX31208-42 MFLX40615-49 MFLX30401-60 MFLX31208-45 MFLX30401-63 MFLX31208-44
Masterflex® Reducer Fittings, Hose Barb, Tee, Avantor®
Fittings and Connectors
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).