Masterflex® Union Fittings, Hose Barb, Elbow, Polypropylene, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
L0-1.5-200PP 10PK L0-1-200PP(10PK) L0-16PP(5PACK) CDA49-PR0 L0-3PP(100 PACK) 227185 L0-2-200HDPE (10 PACK) L0-10PP(100/PK) L0-6PP(10 PACK) L0-1.5-200PP(100 PACK) LO-2-200PP (100 PACK) L0-1-200PP (10 PACK) 227215 CDA73-PR0 L0-8PP(10 PACK) L0-1.5-200PP(10 PACK) MAKE-UP 227165 LO-2-200PP (10 PACK) L0-4PP(100/PK) L0-2-200HDPE (100 PACK) L0-10PP(10 PACK) L0-6 AGPP 227205 227195 L0-3PP(10 PACK) L0-6PP(100/PK) L0-16PP 50/PK L0-12PP(5 PACK) L0-2-200AGPP CDA10-PR0 L0-2.5PP(10PK) 227175 L0-4PP(10 PACK) 227225
MFLX40626-75EA 260.7 CAD
MFLX40626-75 MFLX06365-28 MFLX41518-58 MFLX40622-71 MFLX40622-75 MFLX06365-85 MFLX06368-74 MFLX40640-23 MFLX06285-40 MFLX40640-24 MFLX06285-20 MFLX06285-60 MFLX40626-71 MFLX06365-17 MFLX50626-64 MFLX50622-64 MFLX40640-26 MFLX50623-19 MFLX50623-17 MFLX50627-19 MFLX40640-28 MFLX51518-49 MFLX06285-50 MFLX06285-30 MFLX16368-72 MFLX50611-97 MFLX06285-70 MFLX40610-23 MFLX40610-24 MFLX06285-10 MFLX40610-27 MFLX40610-28 MFLX40610-25 MFLX40610-26
Masterflex® Union Fittings, Hose Barb, Elbow, Polypropylene, Avantor®
Fittings and Connectors
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimize downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!