Masterflex® Direct-Reading Variable-Area Flowmeters for Nitrogen, Acrylic, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
ACRZB0V06 ACRZB0V13 ACRZB0V20 ACRYB0V06 ACRYB0V13 ACRZS0V06 ACRZS0V20 ACRYS0V06 ACRYS0V27 ACRZB1V06 ACRZB1V13 ACRZB1V20 ACRYB1V06 ACRYB1V13 ACRYB1V27 ACRZS1V06 ACRZS1V13 ACRZS1V20 ACRYS1V06 ACRYS1V13 ACRYS1V20 ACRYS1V27 MFLX68560-61 MFLX68560-62 MFLX68560-64 MFLX68560-67 MFLX68560-68 MFLX68560-75
MFLX68560-56EA 109.97 CAD
MFLX68560-56 MFLX68560-57 MFLX68560-58 MFLX68560-59 MFLX68560-60 MFLX68560-63 MFLX68560-65 MFLX68560-66 MFLX68560-69 MFLX68560-70 MFLX68560-71 MFLX68560-72 MFLX68560-73 MFLX68560-74 MFLX68560-76 MFLX68560-77 MFLX68560-78 MFLX68560-79 MFLX68560-80 MFLX68560-81 MFLX68560-82 MFLX68560-83 MFLX68560-61 MFLX68560-62 MFLX68560-64 MFLX68560-67 MFLX68560-68 MFLX68560-75
Masterflex® Direct-Reading Variable-Area Flowmeters for Nitrogen, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for nitrogen. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.