Masterflex® Direct-Reading Variable-Area Flowmeters for Oxygen, Acrylic, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
ACRZB0V07 ACRZB0V21 ACRYB0V07 ACRYB0V21 ACRYB0V28 ACRYS0V14 ACRYS0V21 ACRZB1V07 ACRYB1V07 ACRYB1V14 ACRYB1V21 ACRZS1V07 ACRZS1V14 ACRZS1V21 ACRYS1V07 ACRYS1V14 ACRYS1V21 MFLX68560-29 MFLX68560-32 MFLX68560-37 MFLX68560-38 MFLX68560-41 MFLX68560-43 MFLX68560-44 MFLX68560-48 MFLX68560-55 MFLX68560-36
MFLX68560-28EA 109.49 CAD
MFLX68560-28 MFLX68560-30 MFLX68560-31 MFLX68560-33 MFLX68560-34 MFLX68560-39 MFLX68560-40 MFLX68560-42 MFLX68560-45 MFLX68560-46 MFLX68560-47 MFLX68560-49 MFLX68560-50 MFLX68560-51 MFLX68560-52 MFLX68560-53 MFLX68560-54 MFLX68560-29 MFLX68560-32 MFLX68560-37 MFLX68560-38 MFLX68560-41 MFLX68560-43 MFLX68560-44 MFLX68560-48 MFLX68560-55 MFLX68560-36
Masterflex® Direct-Reading Variable-Area Flowmeters for Oxygen, Acrylic, Avantor®
Flow Meters
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for oxygen. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. Flowmeters feature a stable, easy-to-read float for quick measurement readings.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications.