Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
PMR1-010689 PMR1-013703 PMR1-010692 PMR1-016668 PMR1-010706 PMR1-010713 PMR1-010727 PMR1-010730 PMR1-010735 PMR1-010567 PMR1-010568 PMR1-010570 PMR1-010571 PMR1-010572 PMR1-010579 P13/1-112-19C MFLX32000-00 MFLX32000-02 MFLX32000-08 MFLX32000-18 MFLX32000-22 MFLX32001-06 MFLX32001-08 MFLX32001-10 MFLX32001-14 MFLX32001-16 MFLX32001-18 MFLX32001-24 MFLX32002-08 MFLX32002-10 MFLX32002-12 MFLX32030-00 MFLX32030-10 MFLX32031-15 MFLX32032-00 MFLX32032-08 MFLX32032-09 MFLX32032-10 MFLX32032-14 MFLX32032-15 MFLX32032-16
MFLX32000-04EA 323.9 CAD
MFLX32000-04 MFLX32000-06 MFLX32000-10 MFLX32000-12 MFLX32000-14 MFLX32001-00 MFLX32001-04 MFLX32001-12 MFLX32002-00 MFLX32002-04 MFLX32002-06 MFLX32002-14 MFLX32030-08 MFLX32030-09 MFLX32030-14 MFLX32030-15 MFLX32030-16 MFLX32031-00 MFLX32031-10 MFLX32000-00 MFLX32000-02 MFLX32000-08 MFLX32000-18 MFLX32000-22 MFLX32001-06 MFLX32001-08 MFLX32001-10 MFLX32001-14 MFLX32001-16 MFLX32001-18 MFLX32001-24 MFLX32002-08 MFLX32002-10 MFLX32002-12 MFLX32030-00 MFLX32030-10 MFLX32031-15 MFLX32032-00 MFLX32032-08 MFLX32032-09 MFLX32032-10 MFLX32032-14 MFLX32032-15 MFLX32032-16
Masterflex® Panel-Mount Direct-Read Variable-Area Flowmeters for Air, 150-mm Scale, Avantor®
Flow Meters
No more eye strain—front shield magnifies scale 16 percent for easier readings.
  • Direct-reading flowmeters feature a calibrated scale for a specific medium—no need to convert readings to get flow rate
  • Scale will not wear off—made of fused ceramic for durability
  • Ensure accurate readings - vertical tangential locator with hairline accuracies
  • Aluminum fittings are economical and good for general use with noncorrosive gases and liquid
  • 316 Stainless Steel (SS) fittings withstand higher temperatures and pressures, and features excellent chemical compatibility
This full range of direct-reading flowmeters provide a robust solution for your laboratory that eliminates the need for reading conversions. Valved flowmeters have a valve mounted at the inlet (bottom) of the flowmeter. This arrangement is typically used for positive pressure applications for liquids and gases. The valve can be reconfigured to mount at the outlet (top), often for vacuum applications.
Delivery information: Includes 150-mesh inlet screen.