Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Supplier: VWR International
MFLX33402-04 MFLX33402-10 MFLX33402-12 MFLX33402-14 MFLX33402-34 MFLX33402-38 MFLX33402-40 MFLX33402-44 MFLX33402-48
MFLX33402-04EA 1243.46 CAD
MFLX33402-04 MFLX33402-10 MFLX33402-12 MFLX33402-14 MFLX33402-34 MFLX33402-38 MFLX33402-40 MFLX33402-44 MFLX33402-48
Masterflex® Correlated Variable-Area Flowmeter with High and Low Optical Alarms, Avantor®
Flow Meters
Protect equipment with automatic flow rate monitoring via high and low alarms.
  • Protect your equipment from damage caused by extreme flow rates
  • Features dual-mounted LED receiver/sensors with optical sensors
These 150-mm valved flowmeters feature two noninvasive optical sensors that easily glide up and down both sides of the meter to set high and low flow rates. The meter is ideal for use with an alarm, valve, or other devices to control flow with use of NPN and PNP transistor outputs. The compact design and ease of operation make this meter perfectMore Product Information