Masterflex® Ultra-High Accuracy Turbine Flowmeters/Transmitters, Avantor®
Supplier: Avantor Fluid Handling
Masterflex®
S -111-3-H
MFLX32714-00EA 1224.5 CAD
MFLX32714-00
Masterflex® Ultra-High Accuracy Turbine Flowmeters/Transmitters, Avantor®
Flow Meters
Provides ±0.5% full scale accuracy in a compact, in-line liquid flowmeter.
  • Withstands up to 500 psi
  • Simultaneously displays flow rate
Select from economical Ryton® construction or rugged brass to withstand up to 500 psi. Flowmeters simultaneously display flow rate, plus provide a signal output for datalogging or control. Power your flowmeters with optional AC adapter kits with signal cables, sold separately.
Delivery information: Includes nIST-traceable calibration certificate.