Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Supplier: AALBORG INSTRUMENTS & CONTROL
T51/1-N032-41G T51/1-N032-41S T51/1-N062-01S T51/1-N112-02S T51/1-N082-03S T51/1-N092-04S P01/1-N032-41ST P01/1-N062-01ST P01/1-N082-03G P01/1-N082-03ST P01/1-N092-04ST P01/1-N102-05ST P01/1-N034-39G P01/1-N034-39ST P21/1-N032-41ST P21/1-N062-01ST P21/1-N112-02ST P21/1-N082-03G P21/1-N082-03ST P21/1-N092-04G P21/1-N092-04ST P21/1-N102-05ST P21/1-N034-39G P21/1-N034-39ST P11/1-N062-01ST P11/1-N082-03G P11/1-N082-03ST MFLX03217-58 MFLX03217-63 MFLX03217-68 MFLX03217-74 MFLX03217-76 MFLX03217-79 MFLX03227-02 MFLX03227-04 MFLX03227-08 MFLX03227-12 MFLX03227-14 MFLX03227-20 MFLX03227-24 MFLX03227-84 MFLX03229-05 MFLX03229-09 MFLX03229-13 MFLX03229-25 MFLX03295-00 MFLX03295-02 MFLX03295-04 MFLX03295-06 MFLX03295-08 MFLX03295-12 MFLX03295-14 MFLX03295-20 MFLX03295-22 MFLX03295-24 MFLX03295-26 MFLX03295-28
MFLX03217-52EA 744.18 CAD
MFLX03217-52 MFLX03217-53 MFLX03217-57 MFLX03217-62 MFLX03217-67 MFLX03217-72 MFLX03217-73 MFLX03227-00 MFLX03227-06 MFLX03227-10 MFLX03227-16 MFLX03227-18 MFLX03227-22 MFLX03227-26 MFLX03227-28 MFLX03227-30 MFLX03229-01 MFLX03229-03 MFLX03229-07 MFLX03229-11 MFLX03229-15 MFLX03229-17 MFLX03229-19 MFLX03229-21 MFLX03229-23 MFLX03229-27 MFLX03229-29 MFLX03229-31 MFLX03295-10 MFLX03295-16 MFLX03295-18 MFLX03217-58 MFLX03217-63 MFLX03217-68 MFLX03217-74 MFLX03217-76 MFLX03217-79 MFLX03227-02 MFLX03227-04 MFLX03227-08 MFLX03227-12 MFLX03227-14 MFLX03227-20 MFLX03227-24 MFLX03227-84 MFLX03229-05 MFLX03229-09 MFLX03229-13 MFLX03229-25 MFLX03295-00 MFLX03295-02 MFLX03295-04 MFLX03295-06 MFLX03295-08 MFLX03295-12 MFLX03295-14 MFLX03295-20 MFLX03295-22 MFLX03295-24 MFLX03295-26 MFLX03295-28
Masterflex® Correlated Flowmeters with High-Res Valves, 150-mm Scale, Avantor®
Flow Meters
Don’t sacrifice suitability for precision.
  • Correlated flowmeters feature a generic scale—reference flow curves calibrated to a variety of media, temperatures and pressures to get the flow rate│Use with a variety of media for laboratory and industrial applications
Aluminum is economical and good for general use with noncorrosive gases and liquids. Brass is economical and good for use with water. Stainless steel withstands higher temperature and pressure and features excellent chemical compatibility. Valved meters have valves mounted at the bottom/inlet, typically used for positive pressure with liquids and gases. The valve can be reconfigured to mount at the top/outlet, often for vacuum applications. All correlated flowmeters come with data sheets for water and air at standard temperature and pressure (STP).